accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 1090552-63-8

IUPAC Name: 4-(2-cyclopropyl-1,3-thiazol-4-yl)benzonitrile
Synonyms: 4-(2-cyclopropyl-1,3-thiazol-4-yl)benzonitrile
Molecular Weight (g/mol): 226.3
Molecular Formula: C13H10N2S
InChi Key: SOAYBZVJEBLFEA-UHFFFAOYSA-N
SMILES: N#CC1=CC=C(C=C1)C1=CSC(=N1)C1CC1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results