accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 109227-12-5

IUPAC Name: 2-fluoro-6-(trifluoromethyl)benzoyl chloride
Synonyms: 2-fluoro-6-(trifluoromethyl)benzoyl chloride
Molecular Weight (g/mol): 226.55
Molecular Formula: C8H3ClF4O
InChi Key: ZDRYDSJMVRRAAF-UHFFFAOYSA-N
SMILES: FC1=CC=CC(=C1C(Cl)=O)C(F)(F)F
Percent Purity 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results