accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 109305-98-8

IUPAC Name: 2-bromo-4,5-dimethoxybenzonitrile
Synonyms: 2-bromo-4,5-dimethoxybenzonitrile
Molecular Weight (g/mol): 242.07
Molecular Formula: C9H8BrNO2
InChi Key: BCLBFUAPLHUFET-UHFFFAOYSA-N
SMILES: COC1=C(OC)C=C(C#N)C(Br)=C1
Molecular Weight (g/mol) 
  • (1)
  • (1)
Quantity 
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results