accessibility menu, dialog, popup

UserName

Viewing profile as user:
H3CPCH3H3CH3CHNNN

CAS RN 109348-55-2

IUPAC Name: 2H-1,2,3-benzotriazole; tetrabutylphosphanium
Synonyms: 2H-benzotriazole; tetrabutylphosphonium
Molecular Weight (g/mol): 378.56
Molecular Formula: C22H41N3P
InChi Key: WANXHMXXVLVMSK-UHFFFAOYSA-N
SMILES: N1N=C2C=CC=CC2=N1.CCCC[P+](CCCC)(CCCC)CCCC
Percent Purity 
  • (1)
Physical Form 
  • (1)
Quantity 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results