Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Tetrabutylphosphonium Benzotriazolate 96.0+%, TCI America™
SDP

Catalog No. T200525G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
25 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
T200525G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. T200525G Supplier TCI America Supplier No. T200525G
Add to Cart
Add to Cart

Chemical Identifiers

CAS 109348-55-2
Molecular Formula C22H40N3P
MDL Number MFCD04038148

Specifications

Color Yellow
Quantity 25 g
Formula Weight 377.56
Percent Purity ≥96.0% (T)
Physical Form Crystalline Lumps
Chemical Name or Material Tetrabutylphosphonium Benzotriazolate
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.