accessibility menu, dialog, popup

UserName

Viewing profile as user:
ClONHFF

CAS RN 1094362-94-3

IUPAC Name: 4-(chloromethyl)-N-(3,5-difluorophenyl)benzamide
Synonyms: 4-(chloromethyl)-N-(3,5-difluorophenyl)benzamide
Molecular Weight (g/mol): 281.69
Molecular Formula: C14H10ClF2NO
InChi Key: AQOFMGRCNVYGFX-UHFFFAOYSA-N
SMILES: FC1=CC(NC(=O)C2=CC=C(CCl)C=C2)=CC(F)=C1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results