accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 1098-60-8

IUPAC Name: hydrogen dimethyl({3-[2-(trifluoromethyl)-10H-phenothiazin-10-yl]propyl})amine chloride
Synonyms: triflupromazine hydrochloride triflupromazine hcl triflupromazine monohydrochloride trifluopromazine hydrochloride fluopromazine monohydrochloride
Molecular Weight (g/mol): 388.88
Molecular Formula: C18H20ClF3N2S
InChi Key: FTNWXGFYRHWUKG-UHFFFAOYSA-N
SMILES: [H+].[Cl-].CN(C)CCCN1C2=CC=CC=C2SC2=CC=C(C=C12)C(F)(F)F
Physical Form 
  • (2)
  • (1)
  • (2)
Quantity 
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
Grade 
  • (2)
Percent Purity 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (2)
  • (1)
  • (2)
  • (1)
  • (2)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (2)
  • (1)