Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Triflupromazine, Hydrochloride, MP Biomedicals™

Catalog No. ICN15704480 Shop All MP Biomedicals, Inc Products
Change view
Click to view available options
Quantity:
25 g
100 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
ICN15704480 100 g
ICN15704425 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. ICN15704480 Supplier MP Biomedicals, Inc Supplier No. 0215704480

missing translation for 'validMainframeMsg'

Chemical Identifiers

CAS 1098-60-8
Molecular Formula C18H20ClF3N2S
Molecular Weight (g/mol) 388.88
MDL Number MFCD00058103
InChI Key FTNWXGFYRHWUKG-UHFFFAOYSA-N
Synonym triflupromazine hydrochloride, flumazin, fluorofen, neoprin, nivoman, vesprin hydrochloride, triflupromazine monohydrochloride, triflupromazine hcl, trifluopromazine hydrochloride
PubChem CID 66069
ChEBI CHEBI:9712
IUPAC Name hydrogen dimethyl({3-[2-(trifluoromethyl)-10H-phenothiazin-10-yl]propyl})amine chloride
SMILES [H+].[Cl-].CN(C)CCCN1C2=CC=CC=C2SC2=CC=C(C=C12)C(F)(F)F

Specifications

Quantity 100 g
Formula Weight 388.9
Physical Form Solid
Chemical Name or Material Triflupromazine, Hydrochloride
EINECSNumber 214-149-6
Recommended Storage Store at 2° to 8°C.
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.