accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 1098102-94-3

IUPAC Name: 2,8-bis(octyloxy)-4,10-dithiatricyclo[7.3.0.0³,⁷]dodeca-1,3(7),5,8,11-pentaene
Synonyms: 2,8-bis(octyloxy)-4,10-dithiatricyclo[7.3.0.0³,⁷]dodeca-1,3(7),5,8,11-pentaene
Molecular Weight (g/mol): 446.71
Molecular Formula: C26H38O2S2
InChi Key: LWSGUMOETOXOHL-UHFFFAOYSA-N
SMILES: CCCCCCCCOC1=C2C=CSC2=C(OCCCCCCCC)C2=C1SC=C2
Percent Purity 
  • (1)
Physical Form 
  • (1)
Quantity 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results