accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 109812-64-8

IUPAC Name: 2-methyl-1,5-diphenyl-1H-pyrrole-3-carboxylic acid
Synonyms: 2-methyl-1,5-diphenylpyrrole-3-carboxylic acid
Molecular Weight (g/mol): 277.32
Molecular Formula: C18H15NO2
InChi Key: DWIYTBRYOQDHTE-UHFFFAOYSA-N
SMILES: CC1=C(C=C(N1C1=CC=CC=C1)C1=CC=CC=C1)C(O)=O

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results