Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

2-Methyl-1,5-diphenyl-1H-pyrrole-3-carboxylic acid, 97%, Thermo Scientific™

Catalog No. SP00294DE
Change view
Click to view available options
Quantity:
5 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
SP00294DE 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. SP00294DE Supplier Thermo Scientific Chemicals Supplier No. SP00294DE
Add to Cart

missing translation for 'validMainframeMsg'

Chemical Identifiers

CAS 109812-64-8
Molecular Formula C18H15NO2
Molecular Weight (g/mol) 277.323
MDL Number MFCD01567220
InChI Key DWIYTBRYOQDHTE-UHFFFAOYSA-N
Synonym 2-methyl-1,5-diphenyl-1h-pyrrole-3-carboxylic acid, 1,5-diphenyl-2-methylpyrrole-3-carboxylic acid, 2-methyl-1,5-diphenyl-3-pyrrolecarboxylic acid, 1h-pyrrole-3-carboxylicacid, 2-methyl-1,5-diphenyl, maybridge1_008589, acmc-20mclm, 2-methyl-1,5-diphenyl-pyrrole-3-carboxylic acid
PubChem CID 2735512
IUPAC Name 2-methyl-1,5-diphenylpyrrole-3-carboxylic acid
SMILES CC1=C(C=C(N1C2=CC=CC=C2)C3=CC=CC=C3)C(=O)O

Specifications

CAS Min % 97.0
Assay Percent Range 97%
Quantity 5 g
Formula Weight 277.32
Percent Purity 97%
Chemical Name or Material 2-Methyl-1, 5-diphenyl-1H-pyrrole-3-carboxylic acid
Hazard Statement GHS H Statement:
May cause respiratory irritation.
Causes skin irritation.
Causes serious eye irritation.
Precautionary Statement GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.