accessibility menu, dialog, popup

UserName

Viewing profile as user:
H3CNNOHNONOHOOHO(Z)

CAS RN 109826-27-9

IUPAC Name: (2Z)-but-2-enedioic acid; 1-[1-({7-methyl-8-oxa-2-azatricyclo[8.4.0.0²,⁶]tetradeca-1(14),3,5,10,12-pentaen-7-yl}methyl)piperidin-4-yl]-2,3-dihydro-1H-1,3-benzodiazol-2-one
Synonyms: 1-[1-({7-methyl-8-oxa-2-azatricyclo[8.4.0.0²,⁶]tetradeca-1(14),3,5,10,12-pentaen-7-yl}methyl)piperidin-4-yl]-3H-1,3-benzodiazol-2-one; maleic acid
Molecular Weight (g/mol): 544.61
Molecular Formula: C30H32N4O6
InChi Key: NGODOSILXOFQPH-BTJKTKAUNA-N
SMILES: OC(=O)\C=C/C(O)=O.CC1(CN2CCC(CC2)N2C(=O)NC3=CC=CC=C23)OCC2=CC=CC=C2N2C=CC=C12

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results