Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

CGS 9343B, Tocris Bioscience™
SDP

Catalog No. 225510
Change view
Click to view available options
Quantity:
10 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
22-551-0 10 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. 22-551-0 Supplier R&D Systems Supplier No. 2255/10

Item Discontinued Customers require a valid business account associated to their profile to access pricing and ordering options.

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Calmodulin antagonist

Calmodulin antagonist; inhibits calmodulin-stimulated cAMP phosphodiesterase activity with an IC50 of 3.3 nM. Also prevents estrogen-induced transcription activation by ER, reversibly blocks voltage-activated Na+, Ca2+ and K+ currents in PC12 cells and inhibits nAChR.

Chemical Identifiers

CAS 109826-27-9
Molecular Formula C30H32N4O6
Molecular Weight (g/mol) 544.61
MDL Number MFCD04113002
InChI Key NGODOSILXOFQPH-BTJKTKAUNA-N
Synonym 1,3-dihydro-1-1-4-methyl-4h,6h-pyrrolo 1,2-a 4,1-benzoxazepin-4-yl methyl-4-piperidinyl-2h-benzimidazol-2-one maleate
PubChem CID 14171586
IUPAC Name (2Z)-but-2-enedioic acid; 1-[1-({7-methyl-8-oxa-2-azatricyclo[8.4.0.0²,⁶]tetradeca-1(14),3,5,10,12-pentaen-7-yl}methyl)piperidin-4-yl]-2,3-dihydro-1H-1,3-benzodiazol-2-one
SMILES OC(=O)\C=C/C(O)=O.CC1(CN2CCC(CC2)N2C(=O)NC3=CC=CC=C23)OCC2=CC=CC=C2N2C=CC=C12

Specifications

Quantity 10 mg
Formula Weight 544.61
Percent Purity >98%
Chemical Name or Material CGS 9343B
Recommended Storage Desiccate at Room Temperature
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.