accessibility menu, dialog, popup

UserName

Viewing profile as user:
HN

CAS RN 109871-20-7

IUPAC Name: N-phenylanthracen-2-amine
Synonyms: N-phenylanthracen-2-amine
Molecular Weight (g/mol): 269.35
Molecular Formula: C20H15N
InChi Key: BIWQNAUHSWTLBO-UHFFFAOYSA-N
SMILES: N(C1=CC=CC=C1)C1=CC=C2C=C3C=CC=CC3=CC2=C1
Percent Purity 
  • (1)
Physical Form 
  • (1)
Quantity 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results