accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 1099597-82-6

IUPAC Name: 2-(4-trifluoromethanesulfonylphenyl)acetic acid
Synonyms: (4-trifluoromethanesulfonylphenyl)acetic acid
Molecular Weight (g/mol): 268.21
Molecular Formula: C9H7F3O4S
InChi Key: FIEUCHICQSJBAU-UHFFFAOYSA-N
SMILES: OC(=O)CC1=CC=C(C=C1)S(=O)(=O)C(F)(F)F

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results