accessibility menu, dialog, popup

UserName

Viewing profile as user:
CH3CH3H3COOCH3CH3H3C

CAS RN 110-05-4

IUPAC Name: 2-(tert-butylperoxy)-2-methylpropane
Synonyms: T-butyl-peroxide
Molecular Weight (g/mol): 146.23
Molecular Formula: C8H18O2
InChi Key: LSXWFXONGKSEMY-UHFFFAOYSA-N
SMILES: CC(C)(C)OOC(C)(C)C
Boiling Point 
  • (6)
  • (1)
Molecular Weight (g/mol) 
  • (4)
  • (3)
Quantity 
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
Percent Purity 
  • (1)
  • (3)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (3)
  • (1)
  • (3)
  • (6)
  • (1)
  • (4)
  • (3)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (3)
  • (4)