Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Sigma Aldrich Luperox™ Di, Tert-Butyl Peroxide

Catalog No. 1685215ML Shop All MilliporeSigma Sigma Organic Chemistry Products
Encompass
Encompass_Preferred
Change view
Click to view available options
Quantity:
5 mL
250 mL
1 L
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
16-852-15ML 5 mL
16-852-11L 1 L
16-852-1250ML 250 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Danger
Catalog No. 16-852-15ML Supplier Sigma Aldrich Supplier No. 1685215ML
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Danger

MilliporeSigma Sigma Organics products encompass a wide range of quality reagents, solvents, catalysts, and building blocks for organic synthesis. From benchtop discovery to process development and scale-up, Sigma Organics solutions are built to meet the needs of synthetic chemists.

Luperox™DI, tert-Butyl peroxide has been used as a radical initiator to induce free radical polymerization. It has also been used as a cetane enhancer in a study to determine the phase behavior of carboxylate-based extended surfactant reverse micellar microemulsions with ethanol and vegetable oil/diesel blends.Product of Arkema Inc.

Specifications

CAS 110-05-4
Density 0.796 g/mL at 25°C
Boiling Point 109°C to 110°C
Molecular Formula C8H18O2
Refractive Index n20/D 1.3891
Linear Formula (CH3)3COOC(CH3)3
MDL Number MFCD00008803
Quantity 5 mL
Synonym tert-Butyl peroxide; Di-tert-butyl peroxide
Molecular Weight (g/mol) 146.23 g/mol
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.