accessibility menu, dialog, popup

UserName

Viewing profile as user:
H3CONHHNOCH3

CAS RN 110-30-5

IUPAC Name: N-(2-octadecanamidoethyl)octadecanamide
Synonyms: ethylene distearamide ethylene bis(stearamide)
Molecular Weight (g/mol): 593.04
Molecular Formula: C38H76N2O2
InChi Key: RKISUIUJZGSLEV-UHFFFAOYSA-N
SMILES: CCCCCCCCCCCCCCCCCC(=O)NCCNC(=O)CCCCCCCCCCCCCCCCC
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results