accessibility menu, dialog, popup

UserName

Viewing profile as user:
H3COOOOCH3

CAS RN 110-40-7

IUPAC Name: 1,10-diethyl decanedioate
Synonyms: diethyl sebacate
Molecular Weight (g/mol): 258.36
Molecular Formula: C14H26O4
InChi Key: ONKUXPIBXRRIDU-UHFFFAOYSA-N
SMILES: CCOC(=O)CCCCCCCCC(=O)OCC
Molecular Weight (g/mol) 
  • (1)
  • (3)
Quantity 
  • (1)
  • (1)
  • (1)
  • (1)
Boiling Point 
  • (2)
  • (1)
Percent Purity 
  • (2)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (1)
  • (1)
  • (2)
  • (1)
  • (3)
  • (1)
  • (1)
  • (1)
  • (1)
  • (2)
  • (1)
  • (2)
  • (1)
  • (2)