Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Diethyl Sebacate

Catalog No. ICN211003 Shop All MP Biomedicals, Inc Products
Change view
Click to view available options
Quantity:
100 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
ICN211003 100 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. ICN211003 Supplier MP Biomedicals, Inc Supplier No. 0521100380

missing translation for 'validMainframeMsg'

Chemical Identifiers

CAS 110-40-7
Molecular Formula C14H26O4
Molecular Weight (g/mol) 258.358
InChI Key ONKUXPIBXRRIDU-UHFFFAOYSA-N
Synonym diethyl sebacate, ethyl sebacate, decanedioic acid, diethyl ester, bisoflex des, diethyl 1,10-decanedioate, sebacic acid, diethyl ester, ethyl sebacate van, sebacic acid diethyl ester, diethyl 1,8-octanedicarboxylate, unii-i41b9fjk6v
PubChem CID 8049
IUPAC Name diethyl decanedioate
SMILES CCOC(=O)CCCCCCCCC(=O)OCC

Specifications

Quantity 100 g
Formula Weight 258.36
Chemical Name or Material Diethyl Sebacate
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.