accessibility menu, dialog, popup

UserName

Viewing profile as user:
ClCl(E)

CAS RN 110-57-6

IUPAC Name: (2E)-1,4-dichlorobut-2-ene
Synonyms: 1,4-dichlorobut-2-ene 1,4-dichloro-2-butene(trans)
Molecular Weight (g/mol): 124.99
Molecular Formula: C4H6Cl2
InChi Key: FQDIANVAWVHZIR-OWOJBTEDSA-N
SMILES: ClC\C=C\CCl
Molecular Weight (g/mol) 
  • (2)
  • (3)
Boiling Point 
  • (3)
Percent Purity 
  • (1)
  • (2)
Quantity 
  • (1)
  • (1)
  • (1)
Solvent or Matrix 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (1)
  • (3)
  • (1)
  • (2)
  • (3)
  • (3)
  • (1)
  • (2)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)