Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

trans-1,4-Dichloro-2-butene Solution, 100μg/mL in methanol, Ultra Scientific

Catalog No. USHC4601
Encompass
Change view
Click to view available options
Quantity:
1 x 1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
US-HC-460-1 1 x 1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. US-HC-460-1 Supplier Agilent Technologies Supplier No. HC4601
May include imposed supplier surcharges.
Add to Cart
Add to Cart

Chemical Identifiers

CAS 110-57-6 , 67-56-1
Molecular Formula C4H6Cl2
Molecular Weight (g/mol) 124.992
InChI Key FQDIANVAWVHZIR-OWOJBTEDSA-N
PubChem CID 642197
ChEBI CHEBI:82446
IUPAC Name (E)-1,4-dichlorobut-2-ene
SMILES C(C=CCCl)Cl

Specifications

CAS Max % 99.98, 0.01
Analyte or Component Names Methanol,trans-1,4-dichloro-2-butene
Element Methanol, Trans-1,4-dichloro-2-butene
Solvent or Matrix Methanol
Quantity 1 x 1 mL
Concentration or Composition (by Analyte or Components) 100μg/mL
Chemical Name or Material trans-1, 4-Dichloro-2-butene Solution
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.