accessibility menu, dialog, popup

UserName

Viewing profile as user:
HOOH

CAS RN 110-63-4

IUPAC Name: butane-1,4-diol
Synonyms: 1,4-butanediol 1,4-butylene glycol
Molecular Weight (g/mol): 90.12
Molecular Formula: C4H10O2
InChi Key: WERYXYBDKMZEQL-UHFFFAOYSA-N
SMILES: OCCCCO
Molecular Weight (g/mol) 
  • (5)
  • (5)
Quantity 
  • (2)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
Boiling Point 
  • (2)
  • (5)
  • (2)
Grade 
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (1)
  • (2)
  • (5)
  • (2)
  • (5)
  • (5)
  • (2)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (2)
  • (5)
  • (2)
  • (2)
  • (2)