Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

1,4-Butanediol, BAKER™, J.T. Baker™

Catalog No. 02002013 Shop All Avantor Products
Change view
Click to view available options
Quantity:
500 mL
Packaging:
Narrow Mouth Amber Glass Bottle
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
02-002-013 500 mL Narrow Mouth Amber Glass Bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. 02-002-013 Supplier Avantor Supplier No. D57007
Add to Cart
Add to Cart
This item is not returnable. View return policy

A grade of chemicals for laboratory use

Chemical Identifiers

CAS 110-63-4
Molecular Formula C4H10O2
Molecular Weight (g/mol) 90.12
MDL Number MFCD00002968
InChI Key WERYXYBDKMZEQL-UHFFFAOYSA-N
Synonym 1,4-butanediol, 1,4-butylene glycol, tetramethylene glycol, 1,4-dihydroxybutane, 1,4-tetramethylene glycol, tetramethylene 1,4-diol, sucol b, diol 14b, agrisynth b1d, unii-7xoo2le6g3
PubChem CID 8064
ChEBI CHEBI:41189
IUPAC Name butane-1,4-diol
SMILES OCCCCO

Specifications

Flash Point 121.00°C
Packaging Narrow Mouth Amber Glass Bottle
Quantity 500 mL
Specific Gravity 1L = 1.02kg
Formula Weight 90.12
Chemical Name or Material 1,4-Butanediol
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.