accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 110-68-9

IUPAC Name: butyl(methyl)azanium
Synonyms: butyl(methyl)azanium
Molecular Weight (g/mol): 88.17
Molecular Formula: C5H14N
InChi Key: QCOGKXLOEWLIDC-UHFFFAOYSA-O
SMILES: CCCC[NH2+]C

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results