Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

N-Methylbutylamine 93.0+%, TCI America™
SDP

Catalog No. M017925ML Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
25 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
M017925ML 25 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. M017925ML Supplier TCI America Supplier No. M017925ML
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Chemical Identifiers

CAS 110-68-9
Molecular Formula C5H13N
Molecular Weight (g/mol) 87.166
MDL Number MFCD00009426
InChI Key QCOGKXLOEWLIDC-UHFFFAOYSA-N
Synonym n-methylbutylamine, n-butylmethylamine, methylbutylamine, n-methyl-n-butylamine, butylmethylamine, 1-butanamine, n-methyl, n-methylbutanamine, n-butyl-n-methylamine, n-methyl butylamine, n-methyl-1-butanamine
PubChem CID 8068
ChEBI CHEBI:59019
IUPAC Name N-methylbutan-1-amine
SMILES CCCCNC

Specifications

Color Yellow
Boiling Point 91°C
Quantity 25 mL
UN Number 2945
Formula Weight 87.17
Percent Purity ≥93.0% (GC)
Physical Form Liquid
Chemical Name or Material N-Methylbutylamine
RTECSNumber EO5250000
TSCA Yes
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.