CAS RN 110-83-8
Cyclohexene, 99% stab.
CAS: 110-83-8 Molecular Formula: C6H10 Molecular Weight (g/mol): 82.146 MDL Number: MFCD00001539 InChI Key: HGCIXCUEYOPUTN-UHFFFAOYSA-N Synonym: tetrahydrobenzene,cyclohex-1-ene,benzene tetrahydride,1,2,3,4-tetrahydrobenzene,benzenetetrahydride,hexanaphthylene,1-cyclohexene,cykloheksen,benzene, tetrahydro,3,4,5,6-tetrahydrobenzene PubChem CID: 8079 ChEBI: CHEBI:36404 IUPAC Name: cyclohexene SMILES: C1CCC=CC1
Cyclohexene 99.0+%, TCI America™
CAS: 110-83-8 Molecular Formula: C6H10 Molecular Weight (g/mol): 82.146 MDL Number: MFCD00001539 InChI Key: HGCIXCUEYOPUTN-UHFFFAOYSA-N Synonym: tetrahydrobenzene,cyclohex-1-ene,benzene tetrahydride,1,2,3,4-tetrahydrobenzene,benzenetetrahydride,hexanaphthylene,1-cyclohexene,cykloheksen,benzene, tetrahydro,3,4,5,6-tetrahydrobenzene PubChem CID: 8079 ChEBI: CHEBI:36404 IUPAC Name: cyclohexene SMILES: C1CCC=CC1
Cyclohexene, 99%, pure, stabilized
CAS: 110-83-8 InChI Key: HGCIXCUEYOPUTN-UHFFFAOYSA-N Synonym: tetrahydrobenzene,cyclohex-1-ene,benzene tetrahydride,1,2,3,4-tetrahydrobenzene,benzenetetrahydride,hexanaphthylene,1-cyclohexene,cykloheksen,benzene, tetrahydro,3,4,5,6-tetrahydrobenzene PubChem CID: 8079 ChEBI: CHEBI:36404 IUPAC Name: cyclohexene SMILES: C1CCC=CC1
Cyclohexene, 98%, Spectrum™ Chemical
CAS: 110-83-8 Molecular Weight (g/mol): 82.15
Sigma Aldrich Cyclohexene
MilliporeSigma Sigma Organics products encompass a wide range of quality reagents, solvents, catalysts, and building blocks for organic synthesis. From benchtop discovery to process development and scale-up, Sigma Organics solutions are built to meet the needs of synthetic chemists.
Fisher Science Education™ Cyclohexene
CAS: 110-83-8 Molecular Formula: C6H10 Molecular Weight (g/mol): 82.146 InChI Key: HGCIXCUEYOPUTN-UHFFFAOYSA-N Synonym: tetrahydrobenzene,cyclohex-1-ene,benzene tetrahydride,1,2,3,4-tetrahydrobenzene,benzenetetrahydride,hexanaphthylene,1-cyclohexene,cykloheksen,benzene, tetrahydro,3,4,5,6-tetrahydrobenzene PubChem CID: 8079 ChEBI: CHEBI:36404 IUPAC Name: cyclohexene SMILES: C1CCC=CC1
Cyclohexene (Contains 0.01% tert-Butylcresol as An Inhibitor/Reagent), Fisher Chemical
CAS: 110-83-8 Molecular Formula: C6H10 Molecular Weight (g/mol): 82.146 MDL Number: MFCD00001539 InChI Key: HGCIXCUEYOPUTN-UHFFFAOYSA-N Synonym: tetrahydrobenzene,cyclohex-1-ene,benzene tetrahydride,1,2,3,4-tetrahydrobenzene,benzenetetrahydride,hexanaphthylene,1-cyclohexene,cykloheksen,benzene, tetrahydro,3,4,5,6-tetrahydrobenzene PubChem CID: 8079 ChEBI: CHEBI:36404 IUPAC Name: cyclohexene SMILES: C1CCC=CC1
Methylene Chloride (Pentene Preservative), Fisher BioReagents
CAS: 110-83-8 Molecular Formula: C6H10 Molecular Weight (g/mol): 82.146 InChI Key: HGCIXCUEYOPUTN-UHFFFAOYSA-N Synonym: tetrahydrobenzene,cyclohex-1-ene,benzene tetrahydride,1,2,3,4-tetrahydrobenzene,benzenetetrahydride,hexanaphthylene,1-cyclohexene,cykloheksen,benzene, tetrahydro,3,4,5,6-tetrahydrobenzene PubChem CID: 8079 ChEBI: CHEBI:36404 IUPAC Name: cyclohexene SMILES: C1CCC=CC1
Cyclohexene, SPEX CertiPrep™
CAS: 110-83-8 Molecular Formula: C6H10 Molecular Weight (g/mol): 82.146 InChI Key: HGCIXCUEYOPUTN-UHFFFAOYSA-N PubChem CID: 8079 ChEBI: CHEBI:36404 IUPAC Name: cyclohexene SMILES: C1CCC=CC1