Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Methylene Chloride (Pentene Preservative), Fisher BioReagents

Catalog No. BP11864 Shop All Fisherbrand Products
Change view
Click to view available options
Quantity:
4 L
Packaging:
Glass Bottle
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
BP1186-4 4 L Glass Bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. BP1186-4
Request Bulk or Custom Formats

Item Discontinued This item has been discontinued by the supplier. Please to view product availability in your area.

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy
This solvent is often used in automated synthesis procedures.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 110-83-8 , 513-35-9 , 67-56-1 , 75-09-2
Molecular Formula C6H10
Molecular Weight (g/mol) 82.146
InChI Key HGCIXCUEYOPUTN-UHFFFAOYSA-N
Synonym tetrahydrobenzene, cyclohex-1-ene, benzene tetrahydride, 1,2,3,4-tetrahydrobenzene, benzenetetrahydride, hexanaphthylene, 1-cyclohexene, cykloheksen, benzene, tetrahydro, 3,4,5,6-tetrahydrobenzene
PubChem CID 8079
ChEBI CHEBI:36404
IUPAC Name cyclohexene
SMILES C1CCC=CC1

Specifications

Melting Point -97°C
Absorbance 1 max. at 233nm, 0.12 max. at 240nm, 0.01 max. at 254nm
Color Colorless
Density 1.33g/cm³
Quantity 4 L
Identification Pass Test
Packaging Glass Bottle
Physical Form Liquid
Refractive Index 1.4200 to 1.4230 (at 25°C)
Residue after Evaporation 1ppm max.
Assay Percent Range ≥99.9 %
Titratable Acid 0.0003mEq/g max.
Vapor Pressure 20mmHg at 3502°C
UN Number 1593
Water 25ppm max.
Merck Index 15, 6135
Boiling Point 39°C
Formula Weight 84.93
Percent Purity ≥99.9%
Chemical Name or Material Methylene Chloride, Pentene Preservative
Show More Show Less
Safety handling Solvent Safety handling Check Alert storage code Red
Hazard Statement WARNING!
Precautionary Statement Emergency Overview
Possible cancer hazard. May cause cancer based on animal data. May be harmful if inhaled. May cause irritation of respiratory tract. Inhalation may cause central nervous system effects. WARNING! This product contains a chemical known in the State of California to cause birth defects or. Wear personal protective equipment. Use only under a chemical fume hood. Wash off immediately with plenty of water for at least 15 minutes. Obtain medical attention. Rinse immediately with plenty of water, also under the eyelids, for at least 15 minutes. Obtain medical attention. Do not use mouth-to-mouth resuscitation if victim ingested or inhaled the substance; induce artifici. Do not use mouth-to-mouth resuscitation if victim ingested or inhaled the substance; induce artifici. Get medical attention immediately if symptoms occur.

NFPA

Health:2
Flammability:1
Instability:0
Recommended Storage RT
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.