accessibility menu, dialog, popup

UserName

Viewing profile as user:
ClP

CAS RN 1100-88-5

IUPAC Name: benzyltriphenylphosphanium chloride
Synonyms: benzyltriphenylphosphanium chloride
Molecular Weight (g/mol): 388.87
Molecular Formula: C25H22ClP
InChi Key: USFRYJRPHFMVBZ-UHFFFAOYSA-M
SMILES: [Cl-].C(C1=CC=CC=C1)[P+](C1=CC=CC=C1)(C1=CC=CC=C1)C1=CC=CC=C1
Molecular Weight (g/mol) 
  • (7)
  • (2)
Quantity 
  • (3)
  • (3)
  • (3)
Percent Purity 
  • (2)
  • (5)
Physical Form 
  • (5)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (5)
  • (2)
  • (2)
  • (5)
  • (7)
  • (2)
  • (3)
  • (3)
  • (3)
  • (2)
  • (5)
  • (5)