Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Sigma Aldrich Benzyltriphenylphosphonium Chloride

Catalog No. B3280725G Shop All MilliporeSigma Sigma Organic Chemistry Products
Encompass
Change view
Click to view available options
Quantity:
10 mg
100 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
B3280725G 100 g
B32807100G 10 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. B3280725G Supplier Sigma Aldrich Supplier No. B3280725G
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

MilliporeSigma Sigma Organics products encompass a wide range of quality reagents, solvents, catalysts, and building blocks for organic synthesis. From benchtop discovery to process development and scale-up, Sigma Organics solutions are built to meet the needs of synthetic chemists.

Benzyltriphenylphosphonium Chloride

Specifications

CAS 1100-88-5
Melting Point ≥300°C
Molecular Formula C25H22ClP
Linear Formula C6H5CH2P(Cl)(C6H5)3
Quantity 100 g
MDL Number MFCD00011913
Synonym BTPPC; NSC 116712; Triphenyl(phenylmethyl)phosphonium chloride
Molecular Weight (g/mol) 388.87 g/mol
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.