accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 1101668-39-6

IUPAC Name: 2,5,8,11,14-pentaoxaheptadec-16-yne
Synonyms: 2,5,8,11,14-pentaoxaheptadec-16-yne
Molecular Weight (g/mol): 246.3
Molecular Formula: C12H22O5
InChi Key: QOXHTXZSGABRDF-UHFFFAOYSA-N
SMILES: COCCOCCOCCOCCOCC#C
Percent Purity 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results