accessibility menu, dialog, popup

UserName

Viewing profile as user:
H3CCH3NH3CH3CCH3Cl

CAS RN 11041-12-6

IUPAC Name: 4-[3-(4-ethylphenyl)butyl]-N,N,N-trimethylanilinium chloride
Synonyms: colestyramine 考来烯胺
Molecular Weight (g/mol): 331.93
Molecular Formula: C21H30ClN
InChi Key: KNDHRUPPBXRELB-UHFFFAOYNA-M
SMILES: [Cl-].CCC1=CC=C(C=C1)C(C)CCC1=CC=C(C=C1)[N+](C)(C)C
Grade 
  • (4)
Quantity 
  • (1)
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (4)
  • (4)
  • (4)
  • (4)
  • (1)
  • (1)
  • (1)
  • (1)