accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 110429-35-1

IUPAC Name: (3S,4R)-3-[(2H-1,3-benzodioxol-5-yloxy)methyl]-4-(4-fluorophenyl)piperidine
Synonyms: paroxetine aropax paroxetina paroxetina [inn-spanish] seroxat seroxat CR casbol motivan cis-(-)-paroxetine paroxetine, trans-(+)-
Molecular Weight (g/mol): 329.37
Molecular Formula: C19H20FNO3
InChi Key: AHOUBRCZNHFOSL-YOEHRIQHSA-N
SMILES: FC1=CC=C(C=C1)[C@@H]1CCNC[C@H]1COC1=CC=C2OCOC2=C1
Quantity 
  • (2)
  • (1)
  • (2)
Molecular Weight (g/mol) 
  • (2)
  • (2)
Grade 
  • (1)
  • (1)
Percent Purity 
  • (2)
Physical Form 
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (1)
  • (2)
  • (2)
  • (2)
  • (2)
  • (1)
  • (2)
  • (2)
  • (2)
  • (1)
  • (1)
  • (2)
  • (2)