Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Thermo Scientific Chemicals Paroxetine hydrochloride hemihydrate, 98%

Catalog No. AC462630050
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AC462630050 5 g
AC462630010 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AC462630050 Supplier Thermo Scientific Chemicals Supplier No. 462630050

missing translation for 'validMainframeMsg'

CAS: 110429-35-1 | C19H20FNO3 | 329.37 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 110429-35-1
Molecular Formula C19H20FNO3
Molecular Weight (g/mol) 329.37
InChI Key AHOUBRCZNHFOSL-YOEHRIQHSA-N
IUPAC Name (3S,4R)-3-[(2H-1,3-benzodioxol-5-yloxy)methyl]-4-(4-fluorophenyl)piperidine
SMILES FC1=CC=C(C=C1)[C@@H]1CCNC[C@H]1COC1=CC=C2OCOC2=C1

Specifications

Color Clear Colorless
Assay Percent Range 97.5 % min. (HPLC)
Quantity 5 g
Solubility Information Solubility aq. soln.: very slightly soluble. Other solubilities: soluble in methanol, dmf, ethanol
Formula Weight 374.83
Chemical Name or Material Paroxetine hydrochloride hemihydrate
Hazard Category GHS Signal Word: Warning
Hazard Statement GHS H Statement Harmful if swallowed.

Causes skin irritation.

Causes serious eye irritation.

Precautionary Statement GHS P Statement IF SWALLOWED: rinse mouth.

Do NOT induce vomiting.

Call a POISON CENTER or doctor/physician if you feel unwell.

IF ON SKIN: Wash with plenty of soap and water.

If eye irritation persists: Get medical advice/attention.

Wear protective gloves/protective clothing/eye protection/face protection.

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.