accessibility menu, dialog, popup

UserName

Viewing profile as user:
H3COOOOBr

CAS RN 110429-45-3

IUPAC Name: 13-bromo-2,5,8,11-tetraoxatridecane
Synonyms: 13-bromo-2,5,8,11-tetraoxatridecane
Molecular Weight (g/mol): 271.15
Molecular Formula: C9H19BrO4
InChi Key: YFFFQGXWMHAJPP-UHFFFAOYSA-N
SMILES: COCCOCCOCCOCCBr
Percent Purity 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results