CAS RN 110683-10-8
IUPAC Name:
2-[(2E)-3-(4-pentylphenyl)prop-2-enamido]benzoic acid
Synonyms:
2-[(2E)-3-(4-pentylphenyl)prop-2-enamido]benzoic acid
Molecular Weight (g/mol):
337.42
Molecular Formula:
C21H23NO3
InChi Key:
GAMRBCZMOOMBSQ-CCEZHUSRSA-N
SMILES:
CCCCCC1=CC=C(\C=C\C(=O)NC2=CC=CC=C2C(O)=O)C=C1
1
–
2
of
2
results
1
Promotions Available
Thermo Scientific Chemicals N-(4-n-Pentylcinnamoyl)anthranilic acid
Thermo Scientific Chemicals N-(4-n-Pentylcinnamoyl)anthranilic acid
CAS: 110683-10-8 Molecular Formula: C21H23NO3 Synonym: ACA; N-(p-Amylcinnamoyl)anthranilic acid
2
MilliporeSigma™ Calbiochem™ ACA
A cell-permeable inhibitor of phospholipase A2 that blocks epinephrine-stimulated thromboxane production in platelets.