CAS RN 11070-73-8
IUPAC Name:
—
Synonyms:
—
Molecular Weight (g/mol):
—
Molecular Formula:
—
InChi Key:
—
SMILES:
—
1
–
2
of
2
results
1
CERILLIANT™ Insulin from bovine pancreas Solution, 100 μg/mL (PBS pH 7.2), Sold by MilliporeSigma™ Supelco™
Certified reference material