Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

ProteoMass™ Insulin MALDI-MS Standard, MilliporeSigma™ Supelco™

Catalog No. 111012459 Shop All MilliporeSigma Supelco Products
Change view
Click to view available options
Quantity:
5 x 1 Vial
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
11-101-2459 5 x 1 Vial
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 11-101-2459 Supplier MilliporeSigma Supelco Supplier No. I62795X1VL
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Specifications

Packaging Vial
CAS 11070-73-8
Product Line ProteoMass
Linear Formula C254H377N65O75S6
MDL Number MFCD00131380
Synonym Insulin from bovine pancreas
Type MALDI-MS Standard
Quantity 5 x 1 Vial
Formula Weight 5733.49

ProteoMass is a trademark of Sigma-Aldrich Co. LLC

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.