accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 1107608-80-9

IUPAC Name: 2,3-bis[tert-butyl(methyl)phosphanyl]quinoxaline
Synonyms: 2,3-bis[tert-butyl(methyl)phosphanyl]quinoxaline
Molecular Weight (g/mol): 334.38
Molecular Formula: C18H28N2P2
InChi Key: DRZBLHZZDMCPGX-UHFFFAOYNA-N
SMILES: CP(C1=C(N=C2C=CC=CC2=N1)P(C)C(C)(C)C)C(C)(C)C
Molecular Weight (g/mol) 
  • (1)
  • (1)
Quantity 
  • (2)
Percent Purity 
  • (1)
Physical Form 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (1)
  • (1)
  • (1)
  • (1)
  • (2)
  • (1)
  • (1)