Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Sigma Aldrich (Dimethylphenylsilyl)Boronic Acid Pinacol Ester

Catalog No. 6834345G Shop All MilliporeSigma Sigma Organic Chemistry Products
Encompass
Change view
Click to view available options
Quantity:
100 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
68-343-45G 100 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 68-343-45G Supplier Sigma Aldrich Supplier No. 6834345G
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

MilliporeSigma Sigma Organics products encompass a wide range of quality reagents, solvents, catalysts, and building blocks for organic synthesis. From benchtop discovery to process development and scale-up, Sigma Organics solutions are built to meet the needs of synthetic chemists.

(Dimethylphenylsilyl)boronic acid pinacol ester (Suginome'sas reagent) can be used as a reagent: For the selective addition of dimethylphenylsilanyl group to cyclic and acyclic unsaturated ketones, esters, acrylonitriles using a copper catalyst. In the synthesis of (Z)-4-boryl-1-silyl-2-alkene derivatives by stereoselective addition of silicon-boron bond to acyclic 1,3-dienes in presence of Ni catalyst. In the preparation of silyl-substituted butenoate and +a-silyl-substituted acrylate derivatives from allenes and propiolate derivatives via hydrosilylation reactions using a copper catalyst. In the palladium-catalyzed asymmetric silaboration of allenes and alkanes to offered corresponding +a-borylallylsilanes and 2-boryl-1-silylalkanes respectively.

Specifications

CAS 1107608-80-9
Melting Point 100°C to 105°C
Molecular Formula C18H28N2P2
Linear Formula C18H28N2P2
Refractive Index n20/D 1.4946
MDL Number MFCD10567042
Quantity 100 mg
Synonym 2-(Dimethylphenylsilyl)-4,4,5,5-tetramethyl-1,3,2-dioxaborolane; B-(dimethylphenylsilyl)pinacolborane
Molecular Weight (g/mol) 334.38 g/mol
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.