accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 110993-40-3

IUPAC Name: N-tert-butyl-3,5-dimethylaniline
Synonyms: N-tert-butyl-3,5-dimethylaniline
Molecular Weight (g/mol): 177.29
Molecular Formula: C12H19N
InChi Key: HMWHVIXDKZHDEO-UHFFFAOYSA-N
SMILES: CC1=CC(NC(C)(C)C)=CC(C)=C1
Percent Purity 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results