Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

N-tert-Butyl-3,5-dimethylaniline 98.0+%, TCI America™
SDP

Catalog No. B29035G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
B29035G 5 g
B29031G 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. B29035G Supplier TCI America Supplier No. B29035G
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

CAS: 110993-40-3 | C12H19N | 177.291 g/mol

Chemical Identifiers

CAS 110993-40-3
Molecular Formula C12H19N
Molecular Weight (g/mol) 177.291
InChI Key HMWHVIXDKZHDEO-UHFFFAOYSA-N
Synonym n-tert-butyl-3,5-dimethylaniline, n-tert-butyl-3,5-xylidine, 3,5-dimethyl-n-tert-butylaniline, n-1,1-dimethylethyl-3,5-dimethyl benzenamine, benzenamine,n-1,1-dimethylethyl-3,5-dimethyl
PubChem CID 5242234
IUPAC Name N-tert-butyl-3,5-dimethylaniline
SMILES CC1=CC(=CC(=C1)NC(C)(C)C)C

Specifications

Quantity 5 g
Physical Form Clear Liquid at 20°C
Boiling Point 70°C
Formula Weight 177.29
Percent Purity ≥98.0% (GC)
Grade Not Available
Chemical Name or Material N-tert-Butyl-3,5-dimethylaniline
TSCA Yes
Recommended Storage Refrigerator
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.