CAS RN 111-01-3
Squalane, Tech. 90% (Vegetable Squalane), Thermo Scientific Chemicals
CAS: 111-01-3 Molecular Formula: C30H62 Molecular Weight (g/mol): 422.826 MDL Number: MFCD00008953 InChI Key: PRAKJMSDJKAYCZ-UHFFFAOYSA-N Synonym: squalane,perhydrosqualene,dodecahydrosqualene,cosbiol,vitabiosol,spinacane,robane,hexamethyltetracosane,tetracosane, 2,6,10,15,19,23-hexamethyl,hexamethyl tetracosane PubChem CID: 8089 IUPAC Name: 2,6,10,15,19,23-hexamethyltetracosane SMILES: CC(C)CCCC(C)CCCC(C)CCCCC(C)CCCC(C)CCCC(C)C
Squalane, NF, 97-102%, Spectrum™ Chemical
CAS: 111-01-3 Molecular Formula: C30H62 Molecular Weight (g/mol): 422.83 InChI Key: PRAKJMSDJKAYCZ-UHFFFAOYNA-N IUPAC Name: 2,6,10,15,19,23-hexamethyltetracosane SMILES: CC(C)CCCC(C)CCCC(C)CCCCC(C)CCCC(C)CCCC(C)C
Squalane, Spectrum™ Chemical
CAS: 111-01-3 Molecular Weight (g/mol): 422.81 g/mol
Squalane 98.0+%, TCI America™
CAS: 111-01-3 Molecular Formula: C30H62 Molecular Weight (g/mol): 422.826 MDL Number: MFCD00008953 InChI Key: PRAKJMSDJKAYCZ-UHFFFAOYSA-N Synonym: squalane,perhydrosqualene,dodecahydrosqualene,cosbiol,vitabiosol,spinacane,robane,hexamethyltetracosane,tetracosane, 2,6,10,15,19,23-hexamethyl,hexamethyl tetracosane PubChem CID: 8089 IUPAC Name: 2,6,10,15,19,23-hexamethyltetracosane SMILES: CC(C)CCCC(C)CCCC(C)CCCCC(C)CCCC(C)CCCC(C)C
Medisca Squalane NF
Squalane NF is a colorless, practically odorless transparent oil.
Squalane, ≥99.0% (GC), Solstice
CAS: 111-01-3 Molecular Formula: C30H62 Molecular Weight (g/mol): 422.826 MDL Number: MFCD00008953 InChI Key: PRAKJMSDJKAYCZ-UHFFFAOYSA-N Synonym: squalane,perhydrosqualene,dodecahydrosqualene,cosbiol,vitabiosol,spinacane,robane,hexamethyltetracosane,tetracosane, 2,6,10,15,19,23-hexamethyl,hexamethyl tetracosane PubChem CID: 8089 IUPAC Name: 2,6,10,15,19,23-hexamethyltetracosane SMILES: CC(C)CCCC(C)CCCC(C)CCCCC(C)CCCC(C)CCCC(C)C
Sigma Aldrich Squalane
MilliporeSigma Sigma Organics products encompass a wide range of quality reagents, solvents, catalysts, and building blocks for organic synthesis. From benchtop discovery to process development and scale-up, Sigma Organics solutions are built to meet the needs of synthetic chemists.
Sigma Aldrich Squalane
MilliporeSigma Sigma Organics products encompass a wide range of quality reagents, solvents, catalysts, and building blocks for organic synthesis. From benchtop discovery to process development and scale-up, Sigma Organics solutions are built to meet the needs of synthetic chemists.