Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Squalane, Spectrum™ Chemical
SDP

Catalog No. 18621478 Shop All Spectrum Chemical Mfg. Corp. Products
Change view
Click to view available options
Quantity:
100 mL
20 L
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
18-621-478 20 L
18-621-476 100 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 18-621-478 Supplier Spectrum Chemical Manufacturing Corporation Supplier No. S201820LTBL
Get Up to $300 in Additional Chemicals
View all promotions
Add to Cart
Add to Cart
Request a Quote

missing translation for 'validMainframeMsg'

Chemical Identifiers

CAS 111-01-3
Molecular Weight (g/mol) 422.81 g/mol

Specifications

Melting Point -38°C
Packaging Drum
Quantity 20 L
Percent Purity 99%
Physical Form Liquid
Chemical Name or Material Squalane
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.