accessibility menu, dialog, popup

UserName

Viewing profile as user:
HOSOH

CAS RN 111-48-8

IUPAC Name: 2-[(2-hydroxyethyl)sulfanyl]ethan-1-ol
Synonyms: thiodiglycol
Molecular Weight (g/mol): 122.18
Molecular Formula: C4H10O2S
InChi Key: YODZTKMDCQEPHD-UHFFFAOYSA-N
SMILES: OCCSCCO
Molecular Weight (g/mol) 
  • (4)
  • (1)
Percent Purity 
  • (2)
  • (2)
Boiling Point 
  • (2)
Quantity 
  • (1)
  • (1)
Solvent or Matrix 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (4)
  • (1)
  • (1)
  • (4)
  • (1)
  • (2)
  • (2)
  • (2)
  • (1)
  • (1)
  • (1)