accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 111-57-9

IUPAC Name: N-(2-hydroxyethyl)octadecanamide
Synonyms: stearoyl-ethanolamine stearoyl-EA stearamide mea
Molecular Weight (g/mol): 327.55
Molecular Formula: C20H41NO2
InChi Key: OTGQIQQTPXJQRG-UHFFFAOYSA-N
SMILES: CCCCCCCCCCCCCCCCCC(=O)NCCO

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results