Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Stearic Acid Monoethanolamide, MP Biomedicals

Catalog No. ICN222437 Shop All MP Biomedicals, Inc Products
Change view
Click to view available options
Quantity:
100 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
ICN222437 100 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. ICN222437 Supplier MP Biomedicals, Inc Supplier No. 0522243705
Add to Cart
Add to Cart

Chemical Identifiers

CAS 111-57-9
Molecular Formula C20H41NO2
Molecular Weight (g/mol) 327.553
InChI Key OTGQIQQTPXJQRG-UHFFFAOYSA-N
Synonym n-2-hydroxyethyl octadecanamide, n-stearoylethanolamine, stearoylethanolamide, stearoyl ethanolamide, n-2-hydroxyethyl stearamide, stearamide mea, stearamyl, stearic acid monoethanolamide, stearic monoethanolamide, stearic ethanolamide
PubChem CID 27902
ChEBI CHEBI:85299
IUPAC Name N-(2-hydroxyethyl)octadecanamide
SMILES CCCCCCCCCCCCCCCCCC(=O)NCCO

Specifications

Quantity 100 g
Chemical Name or Material Stearic Acid Monoethanolamide
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.