accessibility menu, dialog, popup

UserName

Viewing profile as user:
H3COH

CAS RN 111-70-6

IUPAC Name: heptan-1-ol
Synonyms: heptanol
Molecular Weight (g/mol): 116.2
Molecular Formula: C7H16O
InChi Key: BBMCTIGTTCKYKF-UHFFFAOYSA-N
SMILES: CCCCCCCO
Molecular Weight (g/mol) 
  • (4)
  • (5)
  • (10)
Quantity 
  • (3)
  • (1)
  • (1)
  • (1)
  • (3)
  • (1)
  • (1)
  • (1)
  • (3)
  • (1)
  • (2)
Percent Purity 
  • (2)
  • (1)
  • (5)
  • (6)
  • (3)
Boiling Point 
  • (2)
  • (5)
  • (4)
  • (3)
  • (2)
Physical Form 
  • (13)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (11)
  • (5)
  • (1)
  • (4)
  • (5)
  • (1)
  • (2)
  • (6)
  • (4)
  • (5)
  • (10)
  • (3)
  • (1)
  • (1)
  • (1)
  • (3)
  • (1)
  • (1)
  • (1)
  • (3)
  • (1)
  • (2)
  • (2)
  • (1)
  • (5)
  • (6)
  • (3)
  • (2)
  • (5)
  • (4)
  • (3)
  • (2)
  • (13)
  • (3)
  • (5)
  • (1)