Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

n-Heptanol, MP Biomedicals

Catalog No. ICN202566 Shop All MP Biomedicals, Inc Products
Change view
Click to view available options
Quantity:
100 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
ICN202566 100 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. ICN202566 Supplier MP Biomedicals, Inc Supplier No. 0520256605

missing translation for 'validMainframeMsg'

Chemical Identifiers

CAS 111-70-6
Molecular Formula C7H16O
Molecular Weight (g/mol) 116.20
MDL Number MFCD00002986
InChI Key BBMCTIGTTCKYKF-UHFFFAOYSA-N
Synonym 1-heptanol, heptyl alcohol, heptanol, n-heptanol, n-heptyl alcohol, enanthic alcohol, gentanol, 1-hydroxyheptane, n-heptan-1-ol, enanthyl alcohol
PubChem CID 8129
IUPAC Name heptan-1-ol
SMILES CCCCCCCO

Specifications

Quantity 100 g
Formula Weight 116.2
Chemical Name or Material n-Heptanol
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.