accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 111-81-9

H3COOCH2

CAS RN 111-81-9

IUPAC Name: methyl undec-10-enoate
Synonyms: methyl undecenoate
Molecular Weight (g/mol): 198.31
Molecular Formula: C12H22O2
InChi Key: KISVAASFGZJBCY-UHFFFAOYSA-N
SMILES: COC(=O)CCCCCCCCC=C
Boiling Point 
  • (1)
  • (3)
Molecular Weight (g/mol) 
  • (1)
  • (3)
Percent Purity 
  • (3)
  • (1)
Quantity 
  • (1)
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (1)
  • (3)
  • (3)
  • (1)
  • (3)
  • (1)
  • (3)
  • (3)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (3)